Få mere at vide
L-Alpha-Methyl DOPA Methyl Ester Hydrochloride, TRC
Artikelnummer. 30473392
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473392 10 g
30476125 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30473392 Leverandør Toronto Research Chemicals Leverandørnr. TRCM30381010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 115054-62-1
Molekylær formel C11 H15 N O4 . Cl H
Molekylvægt (g/mol) 261.7
Synonym L-Tyrosine, 3-hydroxy-α-methyl-, methyl ester, hydrochloride (1:1), L-Tyrosine, 3-hydroxy-α-methyl-, methyl ester, hydrochloride (9CI), L-α-Methyldopa methyl ester hydrochloride, Methyl (2S)-2-Amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate Hydrochloride, Methyldopa Methyl Ester Hydrochloride
IUPAC navn methyl (2S)-2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate;hydrochloride
SMIL Cl.COC(=O)[C@@](C)(N)Cc1ccc(O)c(O)c1
Tekniske data
Kemisk navn eller materiale Methyl (2S)-2-Amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate Hydrochloride
Mængde 10 g
InChI formel InChI=1S/C11H15NO4.ClH/c1-11(12,10(15)16-2)6-7-3-4-8(13)9(14)5-7;/h3-5,13-14H,6,12H2,1-2H3;1H/t11-;/m0./s1
Formel vægt 261.0768