Få mere at vide
L-Allo-isoleucine-d10, TRC
Artikelnummer. 30317568
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30317568 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317568 Leverandør Toronto Research Chemicals Leverandørnr. TRCA54820225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Allo-isoleucine-d10
CAS 202529-06-4
Molekylær formel C6H3D10NO2
Mængde 25 mg
Synonym Alloisoleucine-d10, Alloisoleucine, L-d10, (2S,3R)-2-Amino-3-methylpentanoic Acid-d10, (3R)-LS-Isoleucine-d10, Allo-L-Isoleucine-d10, L(+)-Alloisoleucine-d10, Allo-L-Isoleucine-d10, Threo-L-Isoleucine-d10, L-Norvaline-d10, , 3-Methyl-threo- L-allo-Isoleucine-d10, NSC 206282-d10, 2-Amino-3-methyl-pentanoic Acid-D10, [S-(R*,S*)]-2-Amino-3-methylpentanoic Acid-D10, allo-Isoleucine-10
InChI formel InChI=1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5+/m1/s1/i1D3,2D3,3D2,4D,5D
SMIL OC([C@@]([2H])(N)C([2H])([2H][C]([2H])[2H])C([2H])([2H])C([2H])([2H])[2H])=O
IUPAC navn (2S,3R)-2-amino-2,3,4,4,5,5,5-heptadeuterio-3-(trideuteriomethyl)pentanoic acid
Molekylvægt (g/mol) 141.23
Formel vægt 141.1574