Få mere at vide
L-Alanine-N,N,O-d3, CDN
Artikelnummer. 30313072
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30313072 1 g -
30311044 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313072 Leverandør CDN Leverandørnr. CDND61921G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Alanine-N,N,O-D3
CAS 19470-97-4
Molekylær formel C3 D3 H4 N O2
Mængde 1 g
Synonym L-Alanine-N,N,1-d3 (9 CI, ACI), Alanine-N,N,1-d3, L- (8 CI), L-Alanine-N,N,O-d3, L-Alanine-N,N,O-D3, (2 S)-2-Aminopropanoic-N,N,O-d3 acid
InChI formel InChI=1 S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1/i/hD3
SMIL [2 H]OC(=O)[C@H](C)N([2 H])[2 H]
IUPAC navn deuterio (2 S)-2-(dideuterioamino)propanoate
Molekylvægt (g/mol) 92.1117
Formel vægt 92.0665 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity