missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number p-7080597
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2491-20-5
C4H10ClNO2
139.58
MFCD00063663
IYUKFAFDFHZKPI-DOIAFFBONA-N
l-alanine methyl ester hydrochloride, h-ala-ome.hcl, s-methyl 2-aminopropanoate hydrochloride, methyl l-alaninate hydrochloride, h-ala-ome hcl, alanine methyl ester hydrochloride, methyl 2s-2-aminopropanoate hydrochloride, l-ala-ome hcl, d-alanyl ester hydrochloride, l-alanine methyl ester hcl
2733257
methyl (2S)-2-aminopropanoate;hydrochloride
Cl.COC(=O)[C@H](C)N
This item is not returnable. View return policy
2491-20-5
C4H10ClNO2
139.58
MFCD00063663
IYUKFAFDFHZKPI-DOIAFFBONA-N
l-alanine methyl ester hydrochloride, h-ala-ome.hcl, s-methyl 2-aminopropanoate hydrochloride, methyl l-alaninate hydrochloride, h-ala-ome hcl, alanine methyl ester hydrochloride, methyl 2s-2-aminopropanoate hydrochloride, l-ala-ome hcl, d-alanyl ester hydrochloride, l-alanine methyl ester hcl
2733257
methyl (2S)-2-aminopropanoate;hydrochloride
Cl.COC(=O)[C@H](C)N

L-Alanine methyl ester hydrochloride is used to make in-vivo measurement of glucose and alanine metabolism in studies of patients with diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solubility
Soluble in Water (100 mg/ml).

Notes
Hygroscopic store under inert gas Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 106°C to 110°C
Beilstein 3594033
Solubility Information Soluble in Water (100mg/ml).
Formula Weight 139.58
Percent Purity 99%
Quantity 5 g
Optical Rotation +6.5° (c=10 in methanol)
Sensitivity Hygroscopic
Chemical Name or Material L-Alanine methyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'