Få mere at vide
L-Alanine-d7, TRC
Artikelnummer. 30319320
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30319320 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319320 Leverandør Toronto Research Chemicals Leverandørnr. TRCA4815015MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Alanine-d7
CAS 74280-71-0
Molekylær formel C3 D7 N O2
Mængde 5 mg
Synonym L-Alanine-N,N,1,2,3,3,3-d7 (ACI), L-Alanine-d7 (9CI), Alanine-d7 (6CI), (2S)-2-Aminopropanoic-d7 acid, L-Alanine-N,N,O,2,3,3,3-d7, L-Alanine-D7
InChI formel InChI=1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1/i1D3,2D/hD3
SMIL [2H]OC(=O)[C@@]([2H])(N([2H])[2H])C([2H])([2H])[2H]
IUPAC navn deuterio (2S)-2,3,3,3-tetradeuterio-2-(dideuterioamino)propanoate
Molekylvægt (g/mol) 96.1363
Formel vægt 96.0916