Få mere at vide
L-Alanine-d4, TRC
Artikelnummer. 30312290
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30312290 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312290 Leverandør Toronto Research Chemicals Leverandørnr. TRCA48150210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Alanine d4
CAS 18806-29-6
Molekylær formel C3 2H4 H3 N O2
Mængde 10 mg
Synonym L-Alanine-2,3,3,3-d4 (9CI, ACI), Alanine-2,3,3,3-d4, L- (7CI, 8CI), L-Alanine-d4, L-Alanine-2,3,3,3-D4, (2S)-2-Aminopropanoic-2,3,3,3-d4 acid
InChI formel InChI=1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1/i1D3,2D
SMIL [2H]C([2H])([2H])[C@]([2H])(N)C(=O)O
IUPAC navn (2S)-2-amino-2,3,3,3-tetradeuteriopropanoic acid
Molekylvægt (g/mol) 93.12
Formel vægt 93.0728