Få mere at vide
L-Alanine-3,3,3-d3-N-t-BOC, TRC
Artikelnummer. 30321768
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321768 5 mg
30317841 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321768 Leverandør Toronto Research Chemicals Leverandørnr. TRCA4814425MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(tert-Butoxycarbonyl)-L-alanine-3,3,3-D3
CAS 161602-47-7
Molekylær formel C8 D3 H12 N O4
Mængde 5 mg
Synonym L-Alanine-3,3,3-d3, N-[(1,1-dimethylethoxy)carbonyl]- (9CI, ACI), N-[(1,1-Dimethylethoxy)carbonyl]-L-alanine-3,3,3-d3 (ACI), (S)-2-((tert-Butoxycarbonyl)amino)propanoic-3,3,3-d3 acid, (2S)-2-(tert-Butoxycarbonylamino)propionic-3,3,3-d3 acid, (2S)-2-[(2-Methylpropan-2-yl)oxycarbonylamino]propanoic-3,3,3-d3 acid, N-BOC-L-alanine-3,3,3-d3, N-tert-Butoxycarbonyl-L-alanine-3,3,3-d3, N-(tert-Butoxycarbonyl)-L-alanine-3,3,3-D3, N-tert-Boc-L-alanine-3,3,3-d3, L-Alanine-3,3,3-d3-N-t-BOC
InChI formel InChI=1S/C8H15NO4/c1-5(6(10)11)9-7(12)13-8(2,3)4/h5H,1-4H3,(H,9,12)(H,10,11)/t5-/m0/s1/i1D3
SMIL [2H]C([2H])([2H])[C@H](NC(=O)OC(C)(C)C)C(=O)O
IUPAC navn (2S)-3,3,3-trideuterio-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Molekylvægt (g/mol) 192.2275
Formel vægt 192.1189