Få mere at vide
L-Alanine-3,3,3-d3-N-FMOC, CDN
Artikelnummer. 30327797
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30327797 0.5 g 0.5g
30317333 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327797 Leverandør CDN Leverandørnr. CDND57210.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine-3,3,3-D3
CAS 225101-67-7
Molekylær formel C18 D3 H14 N O4
Mængde 0.5 g
Synonym L-Alanine-3,3,3-d3, N-[(9 H-fluoren-9-ylmethoxy)carbonyl]- (9 CI, ACI), N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine-3,3,3-d3 (ACI), (2 S)-2-[[(9 H-Fluoren-9-ylmethoxy)carbonyl]amino]propanoic-3,3,3-d3 acid, FMOC-L-Alanine-3,3,3-d3, N-(9-Fluorenylmethoxycarbonyl)-L-alanine-3,3,3-d3, N-Fmoc-L-alanine-3,3,3-d3, L-Alanine-3,3,3-d3-N-FMOC
InChI formel InChI=1 S/C18H17NO4/c1-11(17(20)21)19-18(22)23-10-16-14-8-4-2-6-12(14)13-7-3-5-9-15(13)16/h2-9,11,16 H,10H2,1H3,(H,19,22)(H,20,21)/t11-/m0/s1/i1D3
SMIL [2 H]C([2 H])([2 H])[C@H](NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O
IUPAC navn (2 S)-3,3,3-trideuterio-2-(9 H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
Molekylvægt (g/mol) 314.3504
Formel vægt 314.1346 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity