Få mere at vide
L-Alanine-2-d1-N-FMOC, CDN
Artikelnummer. 30327071
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327071 0.25 g
30325337 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327071 Leverandør CDN Leverandørnr. CDND61270.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine-2-D
CAS 225101-64-4
Molekylær formel C18 D H16 N O4
Mængde 0.25 g
Synonym L-Alanine-2-d, N-[(9 H-fluoren-9-ylmethoxy)carbonyl]- (9 CI), N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine-2-D, (2 S)-2-[[(9 H-Fluoren-9-ylmethoxy)carbonyl]amino]propanoic-2-d acid, FMOC-L-Alanine-2-d, N-(9-Fluorenylmethoxycarbonyl)-L-alanine-2-d, N-Fmoc-L-alanine-2-d, L-Alanine-2-d1-N-FMOC
InChI formel InChI=1 S/C18H17NO4/c1-11(17(20)21)19-18(22)23-10-16-14-8-4-2-6-12(14)13-7-3-5-9-15(13)16/h2-9,11,16 H,10H2,1H3,(H,19,22)(H,20,21)/t11-/m0/s1/i11D
SMIL [2 H][C@@](C)(NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O
IUPAC navn (2 S)-2-deuterio-2-(9 H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
Molekylvægt (g/mol) 312.338
Formel vægt 312.122 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity