Få mere at vide
L-Alanine-2,3,3,3-d4-N-t-BOC, CDN
Artikelnummer. 30319656
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319656 0.25 g -
30313315 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319656 Leverandør CDN Leverandørnr. CDND57220.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-tert-Boc-alanine-D4
CAS 714964-61-1
Molekylær formel C8 2H4 H11 N O4
Mængde 0.25 g
InChI formel InChI=1 S/C8H15NO4/c1-5(6(10)11)9-7(12)13-8(2,3)4/h5H,1-4H3,(H,9,12)(H,10,11)/t5-/m0/s1/i1D3,5 D
SMIL [2 H]C([2 H])([2 H])[C@]([2 H])(NC(=O)OC(C)(C)C)C(=O)O
IUPAC navn (2 S)-2,3,3,3-tetradeuterio-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Molekylvægt (g/mol) 193.23
Formel vægt 193.1252 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity