Få mere at vide
L-Alanine-2,3,3,3-d4-N-FMOC, CDN
Artikelnummer. 30322213
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322213 0.5 g 0.5g
30319688 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322213 Leverandør CDN Leverandørnr. CDND57230.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Fmoc-L-alanine-d4
CAS 225101-69-9
Molekylær formel C18 D4 H13 N O4
Mængde 0.5 g
Synonym L-Alanine-2,3,3,3-d4, N-[(9 H-fluoren-9-ylmethoxy)carbonyl]- (9 CI), N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine-2,3,3,3-D4, (2 S)-2-[[(9 H-Fluoren-9-ylmethoxy)carbonyl]amino]propanoic-2,3,3,3-d4 acid, FMOC-L-Alanine-2,3,3,3-d4, N-(9-Fluorenylmethoxycarbonyl)-L-alanine-2,3,3,3-d4, N-Fmoc-L-alanine-2,3,3,3-d4, L-Alanine-2,3,3,3-d4-N-FMOC
InChI formel InChI=1 S/C18H17NO4/c1-11(17(20)21)19-18(22)23-10-16-14-8-4-2-6-12(14)13-7-3-5-9-15(13)16/h2-9,11,16 H,10H2,1H3,(H,19,22)(H,20,21)/t11-/m0/s1/i1D3,11 D
SMIL [2 H]C([2 H])([2 H])[C@]([2 H])(NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O
IUPAC navn (2 S)-2,3,3,3-tetradeuterio-2-(9 H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
Molekylvægt (g/mol) 315.3565
Formel vægt 315.1409 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity