Få mere at vide
Tocris Bioscience™ L-733,060 hydrochloride
GREENER_CHOICE
Artikelnummer. 18044262
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18044262 10 mg
18034372 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18044262 Leverandør Tocris Bioscience™ Leverandørnr. 1145/10
Potent NK1 antagonist
Potent NK1 antagonist (Ki values are 0.08, 0.2 and 93.13 nM for gerbil, human and rat receptors, respectively). Produces anxiolytic-like effects in the gerbil elevated plus-maze. Exhibits antitumor activity in vitro. Also decreases HER2 activity and tumor growth in mice bearing HER2+ or EGFR+ breast tumors. Orally bioavailable and brain penetrant.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 148687-76-7
Molekylær formel C20H20ClF6NO
Tekniske data
Kemisk navn eller materiale L-733, 060 hydrochloride
Mængde 10 mg
Formel vægt 439.83
Renhed >99%
Molekylvægt (g/mol) 439.83