missing translation for 'onlineSavingsMsg'
Learn More
L-701324, MedChemExpress Product Code.: 16487588

L-701324, MedChemExpress

Product Code. p-200096871
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16487588

Brand: Medchem Express HY18698

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress L-701324 is an orally active and long acting anticonvulsant with high affinity and selectivity for the glycine site on the NMDA receptor.

L-701324 is an orally active and long acting anticonvulsant with high affinity and selectivity for the glycine site on the NMDA receptor.

Specifications

Chemical Name or Material L-701324
CAS 142326-59-8
Color White
Purity Grade Notes Research
Molecular Formula C21H14ClNO3
Quantity 10 mg
Solubility Information DMSO : ≥ 34 mg/mL (93.46 mM)
SMILES O=C1NC2=C(C=CC(Cl)=C2)C(O)=C1C3=CC=CC(OC4=CC=CC=C4)=C3
Molecular Weight (g/mol) 363.79
Formula Weight 363.79
Percent Purity 99.62%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
L-701324, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.