Få mere at vide
Tocris Bioscience™ L-371,257
Beskrivelse
Potent, high affinity human oxytocin (OT) receptor antagonist (Ki = 4.6 nM) that displays > 800-fold selectivity over human arginine vasopressin receptors V1a and V2. Antagonizes oxytocin-induced contractions in isolated rat uterine tissue (pA2 = 8.44) and in anesthetised rats following intravenous and intraduodenal administration. Decreases length of U-87MG cell projections induced by retinoic acid (Cat.No.0695). Orally active.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | L-371, 257 |
| CAS | 162042-44-6 |
| Mængde | 10 mg |
| Molekylær formel | C28H33N3O6 |
| Formel vægt | 507.59 |
| Renhed | >99% |
| Molekylvægt (g/mol) | 507.587 |