Få mere at vide
L-3-(4-Hydroxy-3-methoxy-d3-phenyl)alanine, TRC
Artikelnummer. 30475534
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30475534 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30475534 Leverandør Toronto Research Chemicals Leverandørnr. TRCL2112915MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-3-(4-Hydroxy-3-methoxy-d3-phenyl)alanine
CAS 586954-09-8
Molekylær formel C10 2H3 H10 N O4
Mængde 5 mg
Synonym L-Tyrosine, 3-(methoxy-d3)- (9CI), 3-(Methoxy-d3)-L-tyrosine, (2S)-2-Amino-3-[4-hydroxy-3-(trideuteriomethoxy)phenyl]propanoic acid, (2S)-2-Amino-3-[4-hydroxy-3-(methoxy-d3)phenyl]propanoic Acid, 3-Methoxy-L-tyrosine-d3, 3-Methoxy-L-tyrosine-D3 (methoxy-D3), (L)-3-O-(Methyl-d3)dopa
InChI formel InChI=1S/C10H13NO4/c1-15-9-5-6(2-3-8(9)12)4-7(11)10(13)14/h2-3,5,7,12H,4,11H2,1H3,(H,13,14)/t7-/m0/s1/i1D3
SMIL [2H]C([2H])([2H])Oc1cc(C[C@H](N)C(=O)O)ccc1O
IUPAC navn (2S)-2-amino-3-[4-hydroxy-3-(trideuteriomethoxy)phenyl]propanoic acid
Molekylvægt (g/mol) 214.23
Formel vægt 214.1033