missing translation for 'onlineSavingsMsg'
Få mere at vide

(L)-2-Bromo-3-phenylpropionic Acid, TRC

Artikelnummer. 30334279
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30334279 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30334279 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68647025G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Kemisk navn eller materiale (L)-2-Bromo-3-phenylpropionic Acid
CAS 35016-63-8
Molekylær formel C9 H9 Br O2
Mængde 25 g
Synonym Benzenepropanoic acid, α-bromo-, (αS)-, Benzenepropanoic acid, α-bromo-, (S)-, (αS)-α-Bromobenzenepropanoic acid, (S)-α-Bromobenzenepropanoic acid, 3-Phenyl-2(S)-bromopropionic acid, L-α-Bromo-β-phenylpropionic acid, (S)-2-Bromo-3-phenylpropionic Acid
InChI formel InChI=1S/C9H9BrO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6H2,(H,11,12)/t8-/m0/s1
SMIL OC(=O)[C@@H](Br)Cc1ccccc1
IUPAC navn (2S)-2-bromo-3-phenylpropanoic acid
Molekylvægt (g/mol) 229.07
Formel vægt 227.9786
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.