Få mere at vide
L-2-Aminopentanoic-4,4,5,5,5-d5 Acid, CDN
Artikelnummer. 30319658
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319658 0.05 g
30309077 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319658 Leverandør CDN Leverandørnr. CDND65680.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale L-Norvaline-4,4,5,5,5-D5
CAS 1202936-50-2
Molekylær formel C5 D5 H6 N O2
Mængde 0.05 g
Synonym L-Norvaline-4,4,5,5,5-d5, L-Norvaline-d5, (2 S)-2-Aminopentanoic-4,4,5,5,5-d5 acid, (S)-2-Aminopentanoic acid-d5, (S)-2-Aminovaleric acid-d5, L-α-Aminovaleric acid-d5, L-Norvaline-4,4,5,5,5-D5
InChI formel InChI=1 S/C5H11NO2/c1-2-3-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1/i1D3,2D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C[C@H](N)C(=O)O
IUPAC navn (2 S)-2-amino-4,4,5,5,5-pentadeuteriopentanoic acid
Molekylvægt (g/mol) 122.177
Formel vægt 122.11 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity