missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-2-Aminobutyric Acid Methyl Ester Hydrochloride, TRC
Specifications
Specifications
| Kemisk navn eller materiale | L-2-Aminobutyric Acid Methyl Ester Hydrochloride |
| CAS | 56545-22-3 |
| Molekylær formel | C5H12ClNO2 |
| Mængde | 10 g |
| Synonym | (S)-2-Amino-butanoic Acid Methyl Ester Hydrochloride, L-alpha-Aminobutyric Acid Methyl Ester Hydrochloride, (S)-2-Aminobutanoic Acid Methyl Ester Hydrochloride |
| InChI formel | InChI=1S/C5H11NO2.ClH/c1-3-4(6)5(7)8-2;/h4H,3,6H2,1-2H3;1H/t4-;/m0./s1 |
| SMIL | Cl.CC[C@H](N)C(=O)OC |
| IUPAC navn | methyl (2S)-2-aminobutanoate;hydrochloride |
| Molekylvægt (g/mol) | 153.61 |
| Formel vægt | 153.0557 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?