Få mere at vide
L-2-Aminobutyric Acid Methyl Ester Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | L-2-Aminobutyric Acid Methyl Ester Hydrochloride |
| CAS | 56545-22-3 |
| Molekylær formel | C5H12ClNO2 |
| Mængde | 1 g |
| Synonym | (S)-2-Amino-butanoic Acid Methyl Ester Hydrochloride, L-alpha-Aminobutyric Acid Methyl Ester Hydrochloride, (S)-2-Aminobutanoic Acid Methyl Ester Hydrochloride |
| InChI formel | InChI=1S/C5H11NO2.ClH/c1-3-4(6)5(7)8-2;/h4H,3,6H2,1-2H3;1H/t4-;/m0./s1 |
| SMIL | Cl.CC[C@H](N)C(=O)OC |
| IUPAC navn | methyl (2S)-2-aminobutanoate;hydrochloride |
| Molekylvægt (g/mol) | 153.61 |
| Formel vægt | 153.0557 |