Få mere at vide
L-2-Aminobutyric-3,3-d2 Acid, TRC
Artikelnummer. 30325369
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30325369 10 mg
30322242 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325369 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60294110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-2-Aminobutanoic-3,3-D2 Acid
CAS 352431-37-9
Molekylær formel C4 D2 H7 N O2
Mængde 10 mg
Synonym Butanoic-3,3-d2 acid, 2-amino-, (2S)- (9CI), (2S)-2-Aminobutanoic-3,3-d2 acid, L-2-Aminobutyric acid-d2, L-Butyrine-d2, L-Ethylglycine-d2, L-Homoalanine-d2, L-α-Aminobutyric-β,β-d2 acid, (S)-2-Aminobutanoic acid-d2, (S)-2-Aminobutanoic-3,3-D2 acid
InChI formel InChI=1S/C4H9NO2/c1-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/t3-/m0/s1/i2D2
SMIL [2H]C([2H])(C)[C@H](N)C(=O)O
IUPAC navn (2S)-2-amino-3,3-dideuteriobutanoic acid
Molekylvægt (g/mol) 105.132
Formel vægt 105.076