Få mere at vide
L-2-Aminobutyric-3,3-d2 Acid, CDN
Artikelnummer. 30322963
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322963 0.005 g
30309164 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322963 Leverandør CDN Leverandørnr. CDND52440.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-2-Aminobutanoic-3,3-D2 Acid
CAS 352431-37-9
Molekylær formel C4 D2 H7 N O2
Mængde 0.005 g
Synonym Butanoic-3,3-d2 acid, 2-amino-, (2 S)- (9 CI), (2 S)-2-Aminobutanoic-3,3-d2 acid, L-2-Aminobutyric acid-d2, L-Butyrine-d2, L-Ethylglycine-d2, L-Homoalanine-d2, L-α-Aminobutyric-β,β-d2 acid, (S)-2-Aminobutanoic acid-d2, (S)-2-Aminobutanoic-3,3-D2 acid
InChI formel InChI=1 S/C4H9NO2/c1-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/t3-/m0/s1/i2D2
SMIL [2 H]C([2 H])(C)[C@H](N)C(=O)O
IUPAC navn (2 S)-2-amino-3,3-dideuteriobutanoic acid
Molekylvægt (g/mol) 105.132
Formel vægt 105.076 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity