missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tocris Bioscience™ KYP 2047

Beskrivelse
KYP 2047 is a high affinity prolyl oligopeptidase (POP) inhibitor (Ki = 0.023 nM). Clears α-synuclein aggregates induced by oxidative stress in neuronal cells. Inhibits the formation of AcSDKP from its precursor 43-mer thymosin β4 (Tβ4). Induces angiogenesis via POP inhibition. It reduces tau aggregation in tau-transfected HEK-293 cells and N2A cells as well as in human iPSC-derived neurons from patients with frontotemporal dementia. In vivo, treatment with KYP 2047 of transgenic mice with tauopathy, reduces tau aggregation in the brain and cerebrospinal fluid and slows cognitive decline. Anti-angiogenic and neuroprotective. Brain penetrant.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2S)-1-[[(2S)-1-(1-Oxo-4-phenylbutyl)-2-pyrrolidinyl]carbonyl]-2-pyrrolidinecarbonitrile |
| CAS | 796874-99-2 |
| Mængde | 25 mg |
| Molekylær formel | C20H25N3O2 |
| Formel vægt | 339.44 |
| Renhed | 98% |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?