missing translation for 'onlineSavingsMsg'
Få mere at vide

STEMCELL Technologies KU0060648

Artikelnummer. 17110322
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17110322 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17110322 Leverandør STEMCELL Technologies Leverandørnr. 74062

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

NHEJ pathway inhibitor; Inhibits DNA-dependent protein kinase (DNA-PK)

  • KU0060648 is an ATP-competitive and highly potent inhibitor of DNA-dependent protein kinase (DNA-PK), with IC50 values of 19 nM in MCF7 human breast cancer cells and 170 nM in SW620 human colon cancer cells
  • KU0060648 also inhibits phosphoinositide 3-kinase (PI3K) with IC50 values of 39 nM in MCF7 cells and > 10 μM in SW620 cells, which suggests a cell-dependent mechanism (Munck et al.)
  • Reduces the frequency of non-homologous end joining (NHEJ) and increases the efficiency of homology-directed repair (HDR) in CRISPR-Cas9 genome editing (Robert et al.)
  • Inhibits tumor growth and sensitizes human cancer cells to DNA double-strand-break (DSB)-inducing therapies (chemo- or radio-therapy) by inhibiting both DNA-PK and PI3K and preventing the execution of DSB repair (Chen et al.; Dietlein et al.; Munck et al.)

Tekniske data

CAS 881375-00-4
Mængde 10 mg
Molekylær formel C33H34N4O4S
Renhed ≥98%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.