Få mere at vide
KU-0063794, MedChemExpress
Artikelnummer. 16498088
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16498088 10 mg
16494007 50 mg
16477856 100 mg
16420558 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16498088 Leverandør Medchem Express Leverandørnr. HY50710
MedChemExpress KU-0063794 is a potent and specific mTOR inhibitor, inhibiting both the mTORC1 and mTORC2 complexes with IC50s of 10 nM.
KU-0063794 is a potent and specific mTOR inhibitor, inhibiting both the mTORC1 and mTORC2 complexes with IC50s of 10 nM.
Tekniske data
Kemisk navn eller materiale KU-0063794
CAS 938440-64-3
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C25H31N5O4
Mængde 10 mg
Opløselighedsinformation DMSO : 16.67 mg/mL (35.81 mM; Need ultrasonic)
SMIL OCC1=CC(C2=CC=C3C(N=C(N=C3N4CCOCC4)N5C[C@@H](O[C@@H](C5)C)C)=N2)=CC=C1OC
Molekylvægt (g/mol) 465.54
Formel vægt 465.54
Procent renhed 95.84%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease