Få mere at vide
Tocris Bioscience™ KT 182
Beskrivelse
KT 182 is a potent, selective and irreversible α/β-hydrolase domain 6 (ABHD6) inhibitor (IC50 = 1.7 nM). KT 182 exhibits selectively for ABHD6 over other brain and liver serine hydrolases, but does exhibit some activity at carboxylesterase-1. KT 182 inhibits ABHD6 activity in the brain and liver in mice following intraperitoneal administration.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | [4-[3 '-(Hydroxymethyl)[1,1 '-biphenyl]-4-yl]-1 H-1,2,3-triazol-1-yl](2-phenyl-1-piperidinyl)methanone |
| CAS | 1402612-62-7 |
| Mængde | 10 mg |
| Mål | Hydrolase Inhibitors |
| Molekylær formel | C27H26N4O2 |
| Renhed | 0.98 |