missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ KRT33A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122048
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
KRT33A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene is a member of the keratin gene family. It is one of the type I hair keratin genes which are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. There are two isoforms of this protein, encoded by two separate genes, KRTHA3A and KRTHA3B. [provided by RefSeq].
Tekniske data
| KRT33A | |
| Polyclonal | |
| Unconjugated | |
| Krt33a | |
| 2310015J09Rik; HA3I; Ha-3I; hair keratin; Hair keratin, type I Ha3-I; hard keratin, type I,3I; HHA3-I; HKA3A; K33A; Ka27; keratin 33A; keratin 33A, type I; keratin, hair, acidic, 3A; keratin, type I cuticular Ha3-I; Keratin-33A; Krt1-3; krt33a; KRTHA3A; ortholog of keratin, hair, acidic, 3A KRTHA3A; RGD1306249; type I hair keratin KA27 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 3883, 71888 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O76009, Q8K0Y2 | |
| Krt33a | |
| Recombinant protein Keratin 33A. The antigen corresponds to amino acid range 57-363 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion