Få mere at vide
Kresoxim-methyl, TRC
Artikelnummer. 30407546
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30407546 50 mg
1 options
Artikelnummer. 30407546 Leverandør Toronto Research Chemicals Leverandørnr. TRCK65900050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Kresoxim-methyl
CAS 143390-89-0
Molekylær formel C18 H19 N O4
Mængde 50 mg
Synonym Kresoxim Methyl, Benzeneacetic acid, α-(methoxyimino)-2-[(2-methylphenoxy)methyl]-, methyl ester, (αE)-, Benzeneacetic acid, α-(methoxyimino)-2-[(2-methylphenoxy)methyl]-, methyl ester, (E)-, BAS 490-02F, BAS 490F, Candit, Cygnus, Discus, Discus (fungicide), Ergon, Ergon (fungicide), Kresoxim-Me, Kresoxim-methyl, RIL-FA 200, Sovran, Strob 1, Stroby, Stroby DF, Stroby WG
InChI formel InChI=1S/C18H19NO4/c1-13-8-4-7-11-16(13)23-12-14-9-5-6-10-15(14)17(19-22-3)18(20)21-2/h4-11H,12H2,1-3H3/b19-17+
SMIL CO\N=C(\C(=O)OC)/c1ccccc1COc2ccccc2C
IUPAC navn methyl (2E)-2-methoxyimino-2-[2-[(2-methylphenoxy)methyl]phenyl]acetate
Molekylvægt (g/mol) 313.35
Formel vægt 313.1314