Beskrivelse
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Kresoxim-methyl |
| CAS | 143390-89-0 |
| CAS Max % | ≤ 100% |
| Lineær formel | C18H19NO4 |
| Beilstein | 8330581 |
| Farve | White |
| Smeltepunkt | 101°C |
| Kogepunkt | 170°C |
| Synonym | Methyl (E)-α-(methoxyimino)-2-(2-methylphenoxymethyl)phenylacetate; Methyl (E)-methoxyimino[α-(o-tolyloxy)-o-tolyl]acetate |
| Molekylvægt (g/mol) | 313.4 g/mol |
| Vis mere |