Få mere at vide
DWK Life Sciences Kimble™ Kontes™ Squibb Pear-Shaped Separatory Funnels with standard taper Stopper and Autoclavable PTFE Stockcock Plug
Kapacitet (metrisk):
125 mL
1000 mL
Stophane:
2; PTFE
4; PTFE
Mængde:
1 Case
4 Case
Pakningsstørrelse:
Hver
Pakke med 4
Beskrivelse
Kimble Chase KimCote™ plastic-coated glassware provides an added measure of safety in the event of breakage
- Lower stems have inside diameter large enough so that a column of liquid will break with the stopcock closed, thereby giving a more complete separation and eliminating the necessity of emptying the funnel
- Designed from ASTM™ Specifications E1096, Type 4 requirements
Tekniske data
Tekniske data
| Kapacitet (metrisk) | 125 mL |
| Materiale | Borosilicate Glass |
| Certificeringer/compliance | ASTM E1096, Type IV |
| Længde (metrisk) stilk | 60mm |
| Mængde | 4 Case |
| Stophane | 2; PTFE |
| Stopmateriale | Glass |
| Produkttype | Squibb Separatory Funnel |