Få mere at vide
Tocris Bioscience™ Ko 143
GREENER_CHOICE
Artikelnummer. 30001334
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30001334 10 mg
18004752 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30001334 Leverandør Tocris Bioscience™ Leverandørnr. 3241/10
Potent and selective BCRP inhibitor
Potent and selective breast cancer resistance protein multidrug transporter (BCRP) inhibitor (EC90 = 26 nM). Displays > 200-fold selectivity over P-gp and MRP-1 transporters. Increases intracellular drug accumulation and reverses BCRP-mediated multidrug resistance. Inhibits ABCB1 and ABCC1 at higher concentrations. Rapidly metabolized in rat plasma.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 461054-93-3
Molekylær formel C26H35N3O5
Tekniske data
Kemisk navn eller materiale Ko 143
Mængde 10 mg
Formel vægt 469.57
Renhed >99%
Molekylvægt (g/mol) 469.58