Learn More
Klever Klever Excel Narrow Cut
Product Code. 17778987
Change view
Click to view available options
Color:
Blue
Orange
Red
Yellow
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17778987 Red Pack of 50
17758987 Blue Pack of 50
17768987 Orange Pack of 50
17788987 Yellow Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17778987 Supplier Klever Supplier No. KCJ420R
  • ONLINE EXCLUSIVE PRICE
    1964.93 DKK
    1617.87 DKK / Pakke med 50
    Spare 347.06 DKK
    17% Slukket
Forventet forsendelse: 21-12-2026
Recessed blade of the Klever Excel Narrow Cut reduces cut injuries and damaged goods.
  • Cuts boxes, banding, film, tape, and thousands more applications.
  • Multiple position handle; two positions for control and reach.
  • Available in both narrow and wide heads with durable, high quality carbon steel blade.
  • Innovative plastic tape splitter.
  • Advanced Plastic Polymers.
  • Highest Safety Level.
  • Disposable.
Specifications
Product Type Klever Excel Narrow Cutter
Disposable Single-use
Sterility Non-sterile
Color Red