Learn More
Kimberly-Clark Professional™ KleenGuard™ G80 Neoprene/Latex Chemical Resistant Gloves
Catalog Number p-8854001
Click to view available options
Size:
Large
Medium
Small
X-Large
G80 gloves have a natural rubber latex construction covered in neoprene to give a high resistance to acids, alkalis, oils and solvents. Ideal for removing paint, sealants and residues from a surface or degreasing machinery with a strong solvent.
  • Neoprene over natural rubber latex construction with cotton flock lining for comfort and perspiration absorption
  • High resistance to acids, oils, solvents and alkalis
  • Meets the EN 374-AKL standard and has a high level of dexterity, per EN420:2003 standard
  • Not suitable for those with Type 1 latex allergies
TRUSTED_SUSTAINABILITY
Certifications/Compliance (Europe) EN 420:2003, EN 374
Color Blue, Yellow
Latex Free No
Material Neoprene, Latex
Size Small
Thickness (English) 27.5 mil
Thickness (Metric) 0.69 mm
EN ISO 374-1 CHEMICAL LETTER CODES AKL
Glove Cuff Style Straight
Grip Design Textured Fingertips
Liner Material Cotton Flock
Lining Lined
Model 38741
Series Kleenguard™ G80
Length (English) 12 in.
Length (Metric) 30.5 cm