Få mere at vide
Tocris Bioscience™ KJ Pyr 9
High affinity Myc inhibitor
Brand: Tocris Bioscience™ 5306/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
KJ Pyr 9 is a high affinity Myc inhibitor (Kd = 6.5 nM). Disrupts Myc-Max interaction. Inhibits Myc-dependent cancer cell line proliferation in vitro. Inhibits growth of MDA-MB-231 breast cancer cell xenografts in mice. Brain penetrant.Tekniske data
| 4-[2-(2-Furanyl)-6-(4-nitrophenyl)-4-pyridinyl]benzamide | |
| 581073-80-5 | |
| 10 mg |