missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kininogen 1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99365AF594
This item is not returnable.
View return policy
Description
Kininogen 1 Polyclonal specifically detects Kininogen 1 in Human samples. It is validated for ELISA.
Specifications
Kininogen 1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
3827 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 594 | |
Alpha-2-thiol proteinase inhibitor, BDK, BK, bradykinin, Fitzgerald factor, High molecular weight kininogen, HMWK, kininogen-1, Williams-Fitzgerald-Flaujeac factor | |
Produced in rabbits immunized with purified, recombinant Human Kininogen 1 (NP_001095886.1; Gln19-Ser644). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction