Få mere at vide
KIN1408, MedChemExpress
Artikelnummer. 16490448
Encompass_Preferred
Skift visning
Mængde:
1 mg
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16490448 5 mg -
16457766 100 mg -
16437988 10 mg 10mg
16403917 50 mg -
16401909 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16490448 Leverandør Medchem Express Leverandørnr. HY19961
MedChemExpress KIN1408 is an agonist of the RIG-1-like receptor (RLR) pathway and exhibits a broad-spectrum antiviral activity. KIN1408 exhibits activity against HCV, influenza A, dengue virus 2, Ebola, Nipah, and Lassa viruses.
KIN1408 is an agonist of the RIG-1-like receptor (RLR) pathway and exhibits a broad-spectrum antiviral activity. KIN1408 exhibits activity against HCV, influenza A, dengue virus 2, Ebola, Nipah, and Lassa viruses.
Tekniske data
Kemisk navn eller materiale KIN1408
Fysisk form Solid
CAS 1903800-11-2
Farve Light Yellow
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C25H19F2N3O3S
Til brug med (applikation) COVID-19-anti-virus
Opløselighedsinformation DMSO : ≥ 39 mg/mL (81.33 mM)
SMIL COC1=CC=C(SC(NC(C2=CC=C(OC(F)F)C=C2)C3=C(O)C(N=CC=C4)=C4C=C3)=N5)C5=C1
Molekylvægt (g/mol) 479.5
Formel vægt 479.5
Procent renhed 98.78%