missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
KIN1148, MedChemExpress
Description
KIN1148, a small-molecule IRF3 agonist, is a novel influenza vaccine adjuvant found to enhance flu vaccine efficacy.
Specifications
Specifications
| Kemisk navn eller materiale | KIN1148 |
| Fysisk form | Solid |
| CAS | 1428729-56-9 |
| Farve | Yellow |
| Mængde | 1 mg |
| Renhedsgrad noter | Research |
| Molekylær formel | C19H11N3OS2 |
| Til brug med (applikation) | COVID-19-anti-virus |
| Opløselighedsinformation | DMSO : ≥ 100 mg/mL (276.67 mM) |
| SMIL | O=C(NC1=NC2=CC=C3C(N=CS3)=C2S1)C4=CC5=C(C=C4)C=CC=C5 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?