Få mere at vide
DWK Life Sciences Kimble™ KIMAX™ Heavy-Duty Clear Glass Class A Volumetric Flasks with Glass Stoppers
Kapacitet (metrisk):
20 mL
Pakningsstørrelse:
Pakke med 6
Beskrivelse
Large numbers make it easy to identify volume from far away
- Wide Mouth
- Heavy, uniform walls provide superior strength, durability and safety
- Large, permanent, easy-to-read markings
- Flat interior bottoms for stirring with standard stir bars
- Graduated to Class A volumetric tolerances
- Calibrated “To Contain”
- Capacity:10 to 250mL
- Calibrated to meet ASTM™ E288
Tekniske data
Tekniske data
| Lukningsmateriale | Glass |
| Kapacitet (metrisk) | 20 mL |
| Materiale | Borosilicate Glass |
| Højde (engelsk) | 5.11 in. |
| Højde (metrisk) | 13 cm |
| Tolerance | ±0.08 mL |
| Uddannet | Yes |
| Mundstil | Wide |
| Lukkestørrelse | 13 |