Få mere at vide
DWK Life Sciences Kimble™ Kontes™ Filtreringssamlinger: Flasker med sidearm, nr. 2 Slangetilslutning
Description
DWK Life Sciences Kimble Kontes Filtreringsenheder: Kolber med sidearm, nr. 2 slangetilslutning, konnektortyper: nr. 2 slangetilslutning, borosilikatglas, Beskrivelse: Filtreringskolbe
- Borosilicate glass filtration flask with side arm hose connection
- #2 hose connection designed for 0.25 in. (6mm) ID flexible tubing
- 125mL flask features a No. 5 stopper joint; 1000mL, 2000mL, and 4000mL flasks feature a No. 8 stopper joint
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
- Suitable for analysis of particulate or microbiological contamination
- A second flask should be connected between the filtering flask and the vacuum source to prevent accidental entry of filtrate into the vacuum line or pump
Order Info
Inkluderer ikke gummiprop og slange
Specifications
Specifications
| Kapacitet (metrisk) | 125 mL |
| Materiale | Borosilikatglas |
| Inkluderer | Nr. 2 Slangeforbindelse |
| Produkttype | Filtreringskolbe |
| Standard konusled | 5 |
| Autoklaverbar | Autoclavable |
| Engangs | Reusable |
| Uddannet | No |
| Mundstil | Joint |
| Sterilitet | Non-sterile |
| Show More |