Få mere at vide
DWK Life Sciences Kimble™ KIMAX™ Serialiserede/certificerede cylindriske mikrokolber
Kapacitet (metrisk):
50 pmol RNA pr. reaktion
2 mL
5 mL
10 mL
25 mL
Pakningsstørrelse:
Pakke med 6
Beskrivelse
Kalibreret til at indeholde
- Permanently marked with individual serial number
- Individually calibrated and certified against equipment whose calibration is traceable to NIST™
- Cylindrical body for better mixing and draining
- Easier to withdraw samples using pipets
- Wide base provides stability
- Marking spot
- Ground glass stopper
- 1 and 2mL sizes come with standard taper 8 stopper; 10mL size with standard taper 9
Overholdelse
Designet ud fra krav fra ASTM™ E 237, klasse A i serie.
Tekniske data
Tekniske data
| Kalibrering | Calibrated To Contain |
| Lukningsmateriale | Glass |
| Kapacitet (metrisk) | 5 mL |
| Materiale | Borosilicate Glass |
| Højde (engelsk) | 3.9 in. |
| Højde (metrisk) | 9.9 cm |
| Tolerance | ±0.02 mL |
| Uddannet | Yes |
| Lukkestørrelse | 8 |