Learn More
DWK Life Sciences Kimble™ KIMAX™ Serialized/Certified Cylindrical Microflasks
Calibrated To Contain
Brand:  DWK Life Sciences 28017A-10
3814.47 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11848492
  • 5095.00 DKK / Pack of 6
Only null left
Estimated Shipment: 21-11-2025
Description
Calibrated To Contain
  • Permanently marked with individual serial number
  • Individually calibrated and certified against equipment whose calibration is traceable to NIST™
  • Cylindrical body for better mixing and draining
  • Easier to withdraw samples using pipets
  • Wide base provides stability
  • Marking spot
  • Ground glass stopper
  • 1 and 2mL sizes come with standard taper 8 stopper; 10mL size with standard taper 9
stopper
Compliance
Designed from requirements of ASTM™ E 237, Class A serialized.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 mL
SDS
Documents Certificates
Promotions
Promotions
Product Content Correction
Product Title
DWK Life Sciences Kimble™ KIMAX™ Serialized/Certified Cylindrical Microflasks > 10mL; Tol.: ±0.02mL; ST Stopper, No. 9