Få mere at vide
DWK Life Sciences Kimble™ KIMAX™ Flasks with Quick-Release Hose Connector
Kapacitet (metrisk):
250 mL
2000 mL
4 L
Pakningsstørrelse:
Hver
Pakke med 2
Beskrivelse
Allow safe attachment of 0.25 in. I.D. (6mm) tubing
- Tubing remains permanently attached to hose barb and opposite end attaches to flask tubulation with positive threaded seal
- Proper positioning of connector provides greater flask stability as tubing angled downward has less tendency to tilt flask
- Graduated with approximate volume scale in durable white ceramic enamel
- Tubing and stoppers sold separately
Tekniske data
Tekniske data
| Certificeringer/compliance | ASTM E1406, Type II, Class 2 |
| Diameter (engelsk) Ydre | 3.26 in. |
| Kapacitet (metrisk) | 250 mL |
| Materiale | Borosilicate Glass |
| Graduation Range | 50 to 250 mL |
| Gradueringer | 25 mL |
| Højde (engelsk) | 6.4 in. |
| Højde (metrisk) | 16.3 cm |
| Diameter (metrisk) Udvendig | 8.3 cm |
| Lukkestørrelse | 6 |