Learn More
Kimberly-Clark™ Professional KleenGuard™ G10 Powder-Free Blue Nitrile Gloves
Ideal for industrial applications and manufacturing
Brand:  Kimberly-Clark™ Professional 57370
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10004814
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +45 70 27 99 20 for assistance
View alternative products »
Description
Description
Order Info
Packaged 100 gloves per dispenser box (90 for X-Large).
Compliance
Certified for food contact.
Specifications
Specifications
Textured fingertips
Beaded/Rolled
Nitrile
7cm
Powder-free
Non-sterile
9.5 x 2.75 in. (24 x 7cm)
Ambidextrous
X-Small
USDA, FDA
Blue
24cm
Videos