Learn More
Kimberly-Clark Professional™ Procedure Masks
22% lighter and cooler than average procedure mask
Brand:  Kimberly-Clark Professional™ 47080
Additional Details : Weight : 3.40190kg
 View more versions of this product
Product Code. 12942495
  • 1080.00 DKK / Pack of 500
Estimated Shipment
08-03-2021
Description
Description
  • Three-layer, pleat-style mask
  • Ear loops
  • Inner facing is light and breathable to reduce irritation
  • Outer facing resists fluid strike through
Specifications
Specifications
CE
Yes
Blue
White tubular knitted Polyester, Polyurethane ear loops
7 in.
≥96% (BFE), ≥97% (PFE)
Pleated
3-ply construction with white hydro-entangled Polyester binding, fully enclosed soft malleable Aluminum nose piece, Polypropylene, Cellulose, Polyurethane facing components
0.1 (PFE), 3 (BFE)μ
18 cm