Learn More
DWK Life Sciences Kimble™ Kim-Kap™ Disposable Closures
Available in natural (clear) or choice of four colors
Brand:  DWK Life Sciences 73665-13
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10084554
  • 1140.00 DKK / Pack of 1000
Estimated Shipment: 31-10-2023
to see stock.
Add to basket
Description
Description
Available in natural (clear) or choice of four colors
  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable
Caps for 38mm O.D. are designed for baffled culture flasks.
Specifications
Specifications
Closure
Polypropylene
13 mm O.D. plain-end culture tubes
Autoclavable
Blue