Få mere at vide
Ki-67 Polyclonal Antibody, Invitrogen™
Beskrivelse
PA5-16785 targets Ki-67 in immunohistochemistry (paraffin) shows reactivity with Human and Rat samples. The PA5-16785 immunogen is a synthetic peptide from C-terminus of human Ki-67. This antibody was orginally validated as part of a Thermo Scientific Cellomics High Content Screening Kit. The antibody sold separately may have slightly different performance and may need to be further optimized for the best results.
Tekniske data
Tekniske data
| Antigen | Ki-67 |
| Applikationer | Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Koncentration | 0.136 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | TBS with 1% BSA and 0.1% sodium azide; pH 7.6 |
| Gene | Mki67 |
| Genadgangsnr. | P46013 |
| Gene Alias | antigen identified by monoclonal antibody Ki 67; antigen identified by monoclonal antibody Ki-67; Antigen identified by monoclonal antibody Ki-67 homolog; Antigen KI-67; Antigen KI-67 homolog; antigen KI-67; proliferation marker protein Ki-67; antigen KI-67-like; cb31; D630048A14Rik; I79_022666; Ki67; Ki-67; KIA; LOW QUALITY PROTEIN: proliferation marker protein Ki-67; marker of proliferation Ki-67; MIB-; MIB-1; Mki67; PPP1R105; Proliferation marker protein Ki-67; proliferation-related Ki-67 antigen; protein phosphatase 1, regulatory subunit 105; RP11-380J17.2; sb:cb31; si:ch211-250b22.7; unnamed protein product; wu:fa11g09; wu:fb57a07; wu:fi14e05 |
| Gensymboler | Mki67 |
| Vis mere |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.