missing translation for 'onlineSavingsMsg'
Learn More

Ketoprofen EP Impurity G, Mikromol™

Product Code. 17971426
Change view
Click to view available options
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
17971426 25 mg
17983786 100 mg
30458009 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17971426 Supplier Mikromol Supplier No. MM0001.200025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ketoprofen EP Impurity G
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3-[(1RS)-1-Cyanoethyl]benzoic Acid
CAS 5537-71-3
Analyt- eller komponentnavne 3-[(1RS)-1-Cyanoethyl]benzoic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C10 H9 N O2
Certificeringer/compliance ISO 17025
Synonym 3-[(1RS)-1-Cyanoethyl]benzoic Acid, Ketoprofen Imp. G (EP), Ketoprofen Impurity G
InChI formel InChI=1S/C10H9NO2/c1-7(6-11)8-3-2-4-9(5-8)10(12)13/h2-5,7H,1H3,(H,12,13)
SMIL CC(C#N)c1cccc(c1)C(=O)O
IUPAC navn 3-(1-cyanoethyl)benzoic acid
Molekylvægt (g/mol) 175.18
Mængde 25 mg
Formel vægt 175.0633 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.