Få mere at vide
(±)-Ketoprofen-d4 (propionic-d4 acid), CDN
Artikelnummer. 30313548
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313548 0.01 g
30318668 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313548 Leverandør CDN Leverandørnr. CDND68370.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (±)-Ketoprofen propionic-d4 acid
CAS 1219805-29-4
Molekylær formel C16 2H4 H10 O3
Mængde 0.01 g
Synonym (±)-Ketoprofen D4 (propionic D4 acid), 2-(3-Benzoylphenyl)-2,3,3,3-tetradeuterio-propanoic acid
InChI formel InChI=1 S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11 H,1H3,(H,18,19)/i1D3,11 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(C(=O)O)c1cccc(c1)C(=O)c2ccccc2
IUPAC navn 2-(3-benzoylphenyl)-2,3,3,3-tetradeuteriopropanoic acid
Molekylvægt (g/mol) 258.31
Formel vægt 258.1194 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity