Få mere at vide
Ketoconazole EP Impurity E, Mikromol™
Artikelnummer. 30051508
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051508 25 mg
17923676 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051508 Leverandør Mikromol Leverandørnr. MM0145.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ketoconazole EP Impurity E
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale cis-[2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methyl p-Tolylsulfonate
CAS 134071-44-6
Analyt- eller komponentnavne cis-[2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methyl p-Tolylsulfonate
Forsendelsestilstand Room Temperature
Molekylær formel C21 H20 Cl2 N2 O5 S
Certificeringer/compliance ISO 17025
Synonym [(2RS,4SR)-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methyl 4-Methylbenzenesulfonate, Ketoconazole Imp. E (EP), 1,3-Dioxolane-4-methanol, 2-(2,4-dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-, 4-(4-methylbenzenesulfonate), (2R,4R)-rel-, 1,3-Dioxolane-4-methanol, 2-(2,4-dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-, 4-methylbenzenesulfonate (ester), (2R,4R)-rel- (9CI), 1,3-Dioxolane-4-methanol, 2-(2,4-dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-, 4-methylbenzenesulfonate (ester), cis-, Ketoconazole Impurity E
InChI formel InChI=1S/C21H20Cl2N2O5S/c1-15-2-5-18(6-3-15)31(26,27)29-12-17-11-28-21(30-17,13-25-9-8-24-14-25)19-7-4-16(22)10-20(19)23/h2-10,14,17H,11-13H2,1H3/t17-,21+/m1/s1
SMIL Cc1ccc(cc1)S(=O)(=O)OC[C@H]2CO[C@@](Cn3ccnc3)(O2)c4ccc(Cl)cc4Cl
IUPAC navn [(2R,4R)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methyl 4-methylbenzenesulfonate
Molekylvægt (g/mol) 483.36
Mængde 25 mg
Formel vægt 482.047 g/mol