missing translation for 'onlineSavingsMsg'
Få mere at vide

Ketoconazole EP Impurity D, Mikromol™

Artikelnummer. 18012170
Change view
Click to view available options
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
18012170 100 mg
18091800 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18012170 Supplier Mikromol Supplier No. MM0145.04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ketoconazole EP Impurity D
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Deacetyl Ketoconazole
CAS 67914-61-8
Analyt- eller komponentnavne Deacetyl Ketoconazole
Forsendelsestilstand Room Temperature
Molekylær formel C24 H26 Cl2 N4 O3
Certificeringer/compliance ISO 9001
Synonym 1-[4-[[(2RS,4SR)-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine, Ketoconazole Imp. D (EP), 1-[4-[[(2RS,4SR)-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine, Ketoconazole Impurity D
InChI formel InChI=1S/C24H26Cl2N4O3/c25-18-1-6-22(23(26)13-18)24(16-29-10-7-28-17-29)32-15-21(33-24)14-31-20-4-2-19(3-5-20)30-11-8-27-9-12-30/h1-7,10,13,17,21,27H,8-9,11-12,14-16H2/t21-,24-/m0/s1
SMIL Clc1ccc(c(Cl)c1)[C@@]2(Cn3ccnc3)OC[C@H](COc4ccc(cc4)N5CCNCC5)O2
IUPAC navn 1-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine
Molekylvægt (g/mol) 489.39
Mængde 100 mg
Formel vægt 488.1382 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.