Få mere at vide
Ketamine EP Impurity B (Esketamine EP Impurity B), Mikromol™
Artikelnummer. 17983876
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17983876 25 mg
17904006 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17983876 Leverandør Mikromol Leverandørnr. MM0144.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ketamine EP Impurity B (Esketamine EP Impurity B)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2RS)-2-(2-Chlorophenyl)-2-hydroxycyclohexanone
CAS 1823362-29-3
Analyt- eller komponentnavne (2RS)-2-(2-Chlorophenyl)-2-hydroxycyclohexanone
Forsendelsestilstand Room Temperature
Molekylær formel C12 H13 Cl O2
Certificeringer/compliance ISO 17025
Synonym Ketamine Hydrochloride Imp. B (EP), Ketamine Imp. B (EP), (2RS)-2-(2-Chlorophenyl)-2-hydroxycyclohexanone, Ketamine Hydrochloride Impurity B, Ketamine Impurity B
InChI formel InChI=1S/C4H3N5O.H2O/c10-4-2-3(6-1-5-2)7-9-8-4;/h1H,(H2,5,6,7,8,10);1H3
SMIL OC1(CCCCC1=O)c2ccccc2Cl
IUPAC navn 2-(2-chlorophenyl)-2-hydroxycyclohexan-1-one
Molekylvægt (g/mol) 224.68
Mængde 25 mg
Formel vægt 224.0604 g/mol